Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyl Sulfone 99+%, Thermo Scientific™

Catalog No. AC108030250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC108030250 25 g
AC108031000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108030250 Supplier Thermo Scientific Chemicals Supplier No. 108030250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-04-9
Molecular Formula C8H18O2S
Molecular Weight (g/mol) 178.29
InChI Key AIDFJGKWTOULTC-UHFFFAOYSA-N
Synonym dibutyl sulfone, butyl sulfone, butane, 1,1'-sulfonylbis, di-n-butyl sulfone, dibutyl sulphone, n-butyl sulfone, 1-butylsulfonyl butane, butylsulfonyl butane, dibutylsulfone, di-n-butylsulfone
PubChem CID 69014
IUPAC Name 1-butylsulfonylbutane
SMILES CCCCS(=O)(=O)CCCC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 43°C to 46°C
Color Beige
Infrared Spectrum Authentic
Linear Formula [CH3(CH2)3]2SO2
Quantity 25 g
Beilstein 01, 371
Formula Weight 178.29
Percent Purity ≥99.0%
Physical Form Adhering Crystals
Chemical Name or Material n-Butyl sulfone, 99+%
Show More Show Less
EINECSNumber 209-914-6
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.