Learn More
n-Butyl vinyl ether, 98%, stab. with 0.01% KOH
Description
Butyl Vinyl Ether is a reagent used in organic chemical reactions and the preparation of pharmaceutical compounds used in anticancer therapy. It is also used in the synthesis of potent inhibitors for PDE5. It is also used in the synthesis of copolymers and as an acrylic/vinyl acetate resin comonomer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water 3 g/L (20°C). Soluble in chloroform and methanol.
Notes
Store away from air and oxidizing agents may form peroxides. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Donot distill to dryness. Keep it refrigerated. Check the container pressure periodically to prevent peroxide formation.
Specifications
Specifications
| Melting Point | -113°C |
| Density | 0.778 |
| Boiling Point | 92°C to 94°C |
| Flash Point | −12°C (10°F) |
| Refractive Index | 1.401 |
| Linear Formula | CH3(CH2)3OCH=CH2 |
| Quantity | 500 mL |
| UN Number | UN3271 |
| Beilstein | 1560217 |
| Solubility Information | Soluble in water 3g/L (20°C). Soluble in chloroform and methanol. |
| Show More |
Safety and Handling
| Hazard Category | H225-H315-H319-H335 |
| Hazard Statement | GHS H Statement H225-H315-H319-H335 Highly flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S. |
| EINECSNumber | 203-860-7 |
| RTECSNumber | KV5950000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.