Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butylbenzene, 99%

Catalog No. AAA109100B
Click to view available options
Quantity:
50 g
250 g
1000 g

CAS: 104-51-8 | C10H14 | 134.22 g/mol

n-Butylbenzene is an organic solvent that has been used to induce cell death in vitro and for bioconversion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Butylbenzene is an organic solvent that has been used to induce cell death in vitro and for bioconversion.

Solubility
Sparingly soluble in water (0.012g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Storage temperature 2 - 8°C. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-51-8
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00009463
InChI Key OCKPCBLVNKHBMX-UHFFFAOYSA-N
Synonym n-butylbenzene, 1-phenylbutane, benzene, butyl, 1-butylbenzene, butyl-benzene, butane, phenyl, unii-s8xz2901rz, butylbenzene, analytical standard, n-butyl benzene, n4b
PubChem CID 7705
ChEBI CHEBI:44194
IUPAC Name butylbenzene
SMILES CCCCC1=CC=CC=C1

Specifications

Melting Point -88°C
Density 0.86
Boiling Point 181°C to 183°C
Flash Point 59°C (138°F)
Odor Aromatic
Refractive Index 1.489
Quantity 1000 g
UN Number UN2709
Beilstein 1903395
Merck Index 14,1549
Solubility Information Sparingly soluble in water (0.012g/L).
Formula Weight 134.22
Percent Purity 99%
Chemical Name or Material n-Butylbenzene
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYL BENZENES
EINECSNumber 203-209-7
RTECSNumber CY9070000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.