Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Butyldeoxynojirimycin, 98%, Thermo Scientific Chemicals

Catalog No. p-7043257
Click to view available options
Quantity:
1 mg
5 mg

An →→alpha-glucosidase Inhibitor

An --alpha-glucosidase Inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72599-27-0
Molecular Formula C10H21NO4
Molecular Weight (g/mol) 219.28
MDL Number MFCD00272581
InChI Key UQRORFVVSGFNRO-UTINFBMNSA-N
PubChem CID 51634
ChEBI CHEBI:50381
IUPAC Name (2R,3R,4R,5S)-1-butyl-2-(hydroxymethyl)piperidine-3,4,5-triol
SMILES CCCCN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO

Specifications

Type N-Butyldeoxynojirimycin
Physical Form Powder
Melting Point 126°C to 127°C
Color White
Quantity 1 mg
Merck Index 14,6188
Solubility Information Soluble in water at 10mg/mL
Formula Weight 219.28
Percent Purity 98%
Chemical Name or Material N-Butyldeoxynojirimycin
RTECSNumber TN4350150
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.