Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
n-Butyllithium, 2.2M in hexane, Thermo Scientific™
Click to view available options
Quantity:
1 mol
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Specifications
CAS | 109-72-8 |
Flash Point | −21°C (−7°F) |
Odor | Petroleum-like |
Molecular Formula | C4H9Li |
Linear Formula | CH3(CH2)3Li |
Quantity | 1 mol |
MDL Number | MFCD00009414 |
UN Number | UN3394 |
Beilstein | 1209227 |
Sensitivity | Air and moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H225-H250-H260-H304-H314-H335-H336-H361f-H373 |
Hazard Statement | GHS H Statement H225-H250-H260-H252-H304-H361-H373-H314-H336 Highly flammable liquid and vapor. Catches fire spontaneously if exposed to air. In contact with water releases flammable gases which may ignite spontaneously. Self-heating in large quantities may catch fire. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. |
Precautionary Statement | P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE |
EINECSNumber | 203-698-7 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction