Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyllithium, 2.5M in hexanes, packaged in resealable septum cap bottle, Thermo Scientific™

Catalog No. aah666889z
Click to view available options
Quantity:
800 mL

Catalog No. AAH666889Z

Supplier: Thermo Scientific Chemicals H666889Z

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-72-8
Molecular Formula C4H9Li
Molecular Weight (g/mol) 64.06
MDL Number MFCD00009414
InChI Key MZRVEZGGRBJDDB-UHFFFAOYSA-N
Synonym n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc
PubChem CID 61028
IUPAC Name butyllithium
SMILES [Li]CCCC

Specifications

Color Yellow
Density 0.693 g/mL
Packaging ChemSeal™ bottles
Linear Formula CH3(CH2)3Li
Quantity 800 mL
UN Number UN3394
Beilstein 1209227
Sensitivity Air and moisture sensitive
Formula Weight 64.06
Physical Form Liquid
Chemical Name or Material n-Butyllithium
Show More Show Less
Hazard Category H225-H250-H260-H304-H314-H335-H336-H361f-H373
Hazard Statement GHS H Statement
H225-H250-H260-H252-H304-H361-H373-H314-H336
Precautionary Statement P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packaging Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
EINECSNumber 203-698-7
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures; Store under Argon
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.