Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyltrichlorosilane, 97+%

Catalog No. p-7081330
Click to view available options
Quantity:
25 g
100 g

CAS: 7521-80-4 | C4H9Cl3Si | 191.551 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7521-80-4
Molecular Formula C4H9Cl3Si
Molecular Weight (g/mol) 191.551
MDL Number MFCD00013605
InChI Key FQEKAFQSVPLXON-UHFFFAOYSA-N
Synonym silane, butyltrichloro, n-butyltrichlorosilane, trichlorobutylsilane, unii-zq9o7lyo00, zq9o7lyo00, butylsilicon trichloride, n-butyl trichlorosilane, butyl trichlorosilane, hsdb 283, butyl-trichlorosilane
PubChem CID 24142
IUPAC Name butyl(trichloro)silane
SMILES CCCC[Si](Cl)(Cl)Cl

Specifications

Boiling Point 142°C to 143°C
Quantity 25 g
Linear Formula CH3(CH2)3SiCl3
UN Number UN1747
Beilstein 1736032
Flash Point 45°C (113°F)
Formula Weight 191.56
Percent Purity ≥97%
Odor Acrid
Refractive Index 1.437
Sensitivity Moisture sensitive
Density 1.16
Chemical Name or Material n-Butyltrichlorosilane
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: BUTYLTRICHLOROSILANE
EINECSNumber 231-381-3
RTECSNumber VV2080000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.