Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyric Acid, FCC, 99%, Spectrum™ Chemical
SDP

Catalog No. 18604765 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
1 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum

Catalog No. 18-604-765

Supplier: Spectrum Chemical Manufacturing Corporation B230745KGBL

B2307, 107-92-6, C4H8O2

Spectrum™ Chemical n-Butyric Acid, FCC is used as an animal feed supplement due to the ability to reduce pathogenic bacterial colonization and is an approved food flavoring in the EU FLAVIS database (number 08.005). The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 107-92-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00002814
InChI Key FERIUCNNQQJTOY-UHFFFAOYSA-N
IUPAC Name butanoic acid
SMILES CCCC(O)=O

Specifications

CAS Max % 1
Percent Purity 99%
Grade FCC
Quantity 45 kg
Packaging Fiber Drum
Refractive Index 1.397 to 1.399
Specific Gravity 0.953-0.957
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.