Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Cadherin Antibody (017), DyLight 594

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289754DL594
Description
N-Cadherin Monoclonal specifically detects N-Cadherin in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
N-Cadherin | |
Monoclonal | |
DyLight 594 | |
cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin | |
This Recombinant N-Cadherin Antibody (017) was obtained from a rabbit immunized with purified, recombinant Human N-Cadherin (NP_001783.2; Met 1-Ala 724). | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunofluorescence | |
17 | |
50 mM Sodium Borate | |
Rabbit | |
100 kDa | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction, Stem Cells | |
1000 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction