Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Cadherin Antibody (13A9), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 35 publications

$203.02 - $578.99

Specifications

Antigen N-Cadherin
Clone 13A9
Concentration 1 mg/mL
Dilution Western Blot 0.5 ug/ml, Simple Western 1:50, Flow Cytometry, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100, Flow (Intracellular), Immunocytochemistry
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP148309SS
SDP
View Documents
Novus Biologicals
NBP148309SS
0.025 mL
Each for $203.02
Only null left
Add to Cart
 
NBP148309
SDP
View Documents
Novus Biologicals
NBP148309
0.1 mL
Each for $578.99
Only null left
Add to Cart
 
Description

Description

N-Cadherin Monoclonal specifically detects N-Cadherin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), Immunocytochemistry.
Specifications

Specifications

N-Cadherin
1 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction, Stem Cells
P19022
1000
This N-Cadherin Antibody (13A9) was developed against the cytoplasmic domain of human N Cadherin [Swiss-Prot# P19022].
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
13A9
Western Blot 0.5 ug/ml, Simple Western 1:50, Flow Cytometry, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100, Flow (Intracellular), Immunocytochemistry
Monoclonal
Purified
RUO
Human, Mouse, Rat
cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin
CDH2
IgG1
Protein G purified
140 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.