Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Cadherin Antibody (8C11), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$292.00 - $572.50
Specifications
Antigen | N-Cadherin |
---|---|
Clone | 8C11 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25332120
![]() |
Novus Biologicals
NBP25332120UG |
20 μg |
Each for $292.00
|
|
|||||
NB253321100
![]() |
Novus Biologicals
NBP253321100UG |
100 μg |
Each for $572.50
|
|
|||||
NB253321200
![]() |
Novus Biologicals
NBP253321200UG |
200 μg | N/A | N/A | N/A | ||||
Description
N-Cadherin Monoclonal specifically detects N-Cadherin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
N-Cadherin | |
0.2 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
1000 | |
Recombinant human N-cadherin extracellular domain (exact sequence is proprietary) | |
Primary | |
Store at 4C. |
8C11 | |
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin | |
CDH2 | |
IgG1 κ | |
Protein A or G purified | |
135 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title