Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Cadherin Antibody (CDH2/1573), Novus Biologicals™

Mouse Monoclonal Antibody
$292.00 - $572.50
Specifications
| Antigen | N-Cadherin |
|---|---|
| Clone | CDH2/1573 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25332820
![]() |
Novus Biologicals
NBP25332820UG |
20 μg |
Each for $292.00
|
|
|||||
NB253328100
![]() |
Novus Biologicals
NBP253328100UG |
100 μg |
Each for $572.50
|
|
|||||
NB253328200
![]() |
Novus Biologicals
NBP253328200UG |
200 μg | N/A | N/A | N/A | ||||
Description
N-Cadherin Monoclonal specifically detects N-Cadherin in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| N-Cadherin | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 1000 | |
| Recombinant human CDH2 intracellular domain (exact sequence is proprietary) | |
| Primary | |
| Store at 4C. |
| CDH2/1573 | |
| Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin | |
| CDH2 | |
| IgG1 κ | |
| Protein A or G purified | |
| 135 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title