Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human N-Cadherin Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB13881G100UG
This item is not returnable.
View return policy
Description
N-Cadherin Monoclonal specifically detects N-Cadherin in Human samples. It is validated for Flow Cytometry.Specifications
N-Cadherin | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P19022.4 | |
CDH2 | |
Mouse myeloma cell line NS0-derived recombinant mouse N-Cadherin Asp160-Ala724 Accession # P19022.4 | |
RUO | |
1000 | |
Human | |
IgM |
Flow Cytometry | |
691723 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
ACOGS, ARVD14, cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, calcium-dependent adhesion protein, neuronal, CD325, CD325 antigen, CDH2, CDHN, CDw325, NCAD, N-cadherin, NCadherin, N-cadherin 1, Neural cadherin, neural-cadherin | |
Mouse | |
100 μg | |
Primary | |
Detects mouse N-Cadherin in direct ELISAs and Western blots. In direct ELISAs approximately 50% cross-reactivity with recombinant mouse N-Cadherin is observed, and no cross-reactivity with recombinant human (rh) E-Cadherin, rhP-Cadherin, rhVE-Cadherin, rhCadherin-4, -8, -11, -12, or -13 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction