Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ N-cadherin, Mouse, Unlabeled, Clone: 6A9.2,

Catalog No. MABT530MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABT530MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT530MI Supplier MilliporeSigma™ Supplier No. MABT530
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cadherin-2 (UniProt P19022; also known as CD325, CDw325, N-cadherin, Neural cadherin) is encoded by the CDH2 (also known as CDHN, NCAD) gene (Gene ID 1000) in human. N-Cadherin is a single-pass type I membrane protein that plays a role in calcium-dependent cell adhesion. N-cadherins preferentially interacts with themselves in a homophilic manner in connecting cells; may thus contribute to the sorting of heterogeneous cell types. Cadherins are synthesized as precursor proteins that are proteolytically cleaved to generate functional, mature proteins. Newly synthesized proN-cadherin (a.a. 1-906) is phosphorylated and proteolytically processed prior to transport to the plasma membrane. Cadherins are composed of an extracellular domain (a.a. 160-724 of human N-cadherin) with five homologous repeats that mediates adhesion, a single pass transmembrane domain (a.a. 725-745 of human N-cadherin), and a conserved cytoplasmic domain (a.a. 746-906 of human N-cadherin) that interacts with catenins to link cadherins to the actin cytoskeleton. N-cadherin is well expressed in endothelial cells and plays an essential role in the maturation and stabilization of normal vessels. However, it also promotes tumor cell survival, migration and invasion, and a high level of its expression is often associated with poor prognosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen N-cadherin
Applications Western Blot
Classification Monoclonal
Clone 6A9.2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene CDH2, CDHN, NCAD
Gene Accession No. P19022
Host Species Mouse
Immunogen GST/His-tagged recombinant human N-cadherin extracellular cadherin 1 domain fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001783
Test Specificity Clone 6A9.2 targets an epitope within N-cadherin extracellular cadherin 1 domain.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.