Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Cadherin Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $524.00
Specifications
Antigen | N-Cadherin |
---|---|
Applications | Western Blot, Immunoprecipitation, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
N-Cadherin Polyclonal specifically detects N-Cadherin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation, Knockout Validated.Specifications
N-Cadherin | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin | |
A synthetic peptide corresponding to a sequence within amino acids 750-850 of human N-Cadherin (NP_001783.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunoprecipitation, KnockDown | |
Unconjugated | |
Rabbit | |
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
PBS with 50% glycerol, pH7.3. | |
1000 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title