Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ N-cadherin Recombinant Rabbit Monoclonal Antibody (020), FITC
GREENER_CHOICE

Catalog No. PIMA546719
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546719 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA546719 Supplier Invitrogen™ Supplier No. MA546719
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human N-cadherin (Met1-Ala724) and conjugated with FITC under optimum conditions, the unreacted FITC was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.

N-cadherin is a 140 kDa protein belonging to a family of transmembrane molecules that mediate calcium-dependent intercellular adhesion. Cadherins are involved in controlling morphogenetic movements during development and regulate cell surface adhesion through homotypic adhesion with the same cadherin species. N-cadherin's function is dependent on its association with the actin-cytoskeleton and is mediated through interactions between the C-terminal region of N-cadherin and the cytoplasmic catenin proteins. The stability of this association is regulated by phosphorylation and dephosphorylation of beta-catenin. Further, N-cadherin is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. The protein functions during gastrulation and is required for establishment of left-right asymmetry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen N-cadherin
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 020
Concentration 100 μg/mL
Conjugate FITC
Formulation PBS with 0.5% BSA and 0.03% ProClin 300
Gene CDH2
Gene Accession No. P19022
Gene Alias bib; cadherin 2; cadherin 2 L homeolog; cadherin 2 type 1 N-cadherin (neuronal); cadherin 2, neuronal; cadherin 2, type 1 preproprotein; cadherin 2, type 1, N-cadherin (neuronal); cadherin 2, type 1, N-cadherin (neuronal) L homeolog; cadherin 2, type 1, N-cadherin a (neuronal); Cadherin2; cadherin-2; Cadherin-2A; Cadherin-N; calcium-dependent adhesion protein, neuronal; CD325; CDH2; cdh2.L; cdh2a; cdh2-a; cdh2-b; CDHN; cdhn-a; CDw325; CDw325 antigen; cell adhesion molecule; glass onion; glo; I79_014914; labyrinth; lyr; NCAD; N-Cad; N-cadherin; N-cadherin 1; N-cadherin A; N-cadherin precursor; N-cadherin-2; neural cadherin; Neural cadherin A; neural cadherin-2; neural calcium-dependent cell adhesion molecule; neural-cadherin; pac; Parachute; unnamed protein product; wu:fb47h04; XELAEV_18031953mg; ZNCAD
Gene Symbols CDH2
Host Species Rabbit
Immunogen Recombinant protein ecoding Human N-cadherin (Met1-Ala724).
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1000
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.