Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-1,2-diaminoethane Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. C151125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C151125G 25 g
C15111G 1 g
C15115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C151125G Supplier TCI America Supplier No. C151125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18807-71-1
Molecular Formula C10H15ClN2O2
Molecular Weight (g/mol) 230.69
MDL Number MFCD00270150
InChI Key QMLKQXIAPAAIEJ-UHFFFAOYSA-N
Synonym benzyl n-2-aminoethyl carbamate hydrochloride, n-cbz-ethylenediamine hydrochloride, benzyl 2-aminoethyl carbamate hydrochloride, n-carbobenzoxy-1,2-diaminoethane hydrochloride, n-z-ethylenediamine hydrochloride, n-benzyloxycarbonyl-1,2-diaminoethane hydrochloride, n-z-1,2-diaminoethane hydrochloride, n-cbz-1,2-diaminoethane hydrochloride, benzyl 2-aminoethyl carbamate hcl, n-benzyloxycarbonylethylenediamine hydrochloride
PubChem CID 12715871
IUPAC Name benzyl N-(2-aminoethyl)carbamate hydrochloride
SMILES Cl.NCCNC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 172°C
Quantity 25 g
Formula Weight 230.69
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-1,2-diaminoethane Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.