Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-4-hydroxy-D-2-phenylglycine 98.0+%, TCI America™
SDP

Catalog No. C277325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C277325G 25 g
C27735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C277325G Supplier TCI America Supplier No. C277325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26787-75-7
Molecular Formula C16H15NO5
Molecular Weight (g/mol) 301.30
MDL Number MFCD09756650
InChI Key CGDQNVNWDDEJPY-UHFFFAOYNA-N
Synonym (R)-2-(Carbobenzoxy)amino-2-(4-hydroxyphenyl)acetic Acid, Z-D-Phg(4-OH)-OH
PubChem CID 1514128
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}-2-(4-hydroxyphenyl)acetic acid
SMILES OC(=O)C(NC(=O)OCC1=CC=CC=C1)C1=CC=C(O)C=C1

Specifications

Color White
Quantity 25 g
Formula Weight 301.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-4-hydroxy-D-2-phenylglycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.