Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-D-glutamic Acid 98.0+%, TCI America™
SDP

Catalog No. C066325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C066325G 25 g
C06635G 5 g
C0663100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C066325G Supplier TCI America Supplier No. C066325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 63648-73-7
Molecular Formula C13H15NO6
Molecular Weight (g/mol) 281.264
MDL Number MFCD00063193
InChI Key PVFCXMDXBIEMQG-SNVBAGLBSA-N
Synonym z-d-glu-oh, z-d-glutamic acid, n-carbobenzoxy-d-glutamic acid, z-d-glu, n-cbz-d-glutamic acid, n-benzyloxycarbonyl-d-glutamic acid, r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-glu-oh, 2r-2-benzyloxy carbonyl amino pentanedioic acid, cbz-d-glu
PubChem CID 1607568
IUPAC Name (2R)-2-(phenylmethoxycarbonylamino)pentanedioic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CCC(=O)O)C(=O)O

Specifications

Melting Point 120°C
Color White
Quantity 25 g
Formula Weight 281.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-D-glutamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.