Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-DL-valine 99.0+%, TCI America™
SDP

Catalog No. C063410G
Click to view available options
Quantity:
1 g
10 g

Chemical Identifiers

CAS 3588-63-4
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD00065127
InChI Key CANZBRDGRHNSGZ-UHFFFAOYSA-N
Synonym n-carbobenzoxy-dl-valine, z-dl-val-oh, n-cbz-dl-valine, 2-benzyloxy carbonyl amino-3-methylbutanoic acid, n-carbobenzoyl-dl-valine, n-benzyloxy carbonyl valine, n-benzyloxycarbonyl-dl-valine, n-carbobenzyl-oxy-dl-valine, dl-valine, n-phenylmethoxy carbonyl
PubChem CID 97743
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoic acid
SMILES CC(C)C(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 78°C
Color White
Quantity 10 g
Formula Weight 251.28
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-DL-valine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.