Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzyloxy-L-glutamic acid 1-methyl ester, 98%, Thermo Scientific™

Catalog No. AC320520010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5672-83-3
Molecular Formula C14H17NO6
Molecular Weight (g/mol) 295.291
MDL Number MFCD00083278
InChI Key BGMCTGARFXPQML-NSHDSACASA-N
Synonym z-glu-ome, z-l-glu-ome, 1-methyl n-carbobenzoxy-l-glutamate, 4s-4-benzyloxy carbonyl amino-5-methoxy-5-oxopentanoic acid, cbz-glu-ome, l-glutamic acid,n-phenylmethoxy carbonyl-, 1-methyl ester, n-cbz-l-glu-ome, z-l-glutamic acid 1-methyl ester, n-cbz-l-glutamic acid 1-methyl ester, n-carbobenzoxy-l-glutamic acid 1-methyl ester
PubChem CID 2733408
IUPAC Name (4S)-5-methoxy-5-oxo-4-(phenylmethoxycarbonylamino)pentanoic acid
SMILES COC(=O)C(CCC(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO2CCH2CH2CH(NHCO2CH2C6H5)CO2CH3
Quantity 1 g
Specific Rotation -21° to -25° (20°C, 589nm) (c=2, C2H5OH)
Formula Weight 295.29
Percent Purity ≥97.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material N-Carbobenzyloxy-L-glutamic acid 1-methyl ester, 98%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.