Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-beta-Methylstyrene, 97%, stabilized

Catalog No. AC150150010
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 873-66-5 | C9H10 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-66-5
Molecular Formula C9H10
Molecular Weight (g/mol) 118.18
MDL Number MFCD00009280
InChI Key QROGIFZRVHSFLM-QHHAFSJGSA-N
Synonym beta-methylstyrene, trans-beta-methylstyrene, 1-phenylpropene, isoallylbenzene, propenylbenzene, trans-1-phenyl-1-propene, 1-propenylbenzene, trans-propenylbenzene, 1-phenyl-1-propene, benzene, 1-propenyl
PubChem CID 252325
IUPAC Name [(E)-prop-1-enyl]benzene
SMILES CC=CC1=CC=CC=C1

Specifications

Color Colorless to Yellow
Density 0.9110g/mL
Boiling Point 175°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5480 to 1.55
Linear Formula C6H5CH=CHCH3
Quantity 1 g
Beilstein 05, 481
Specific Gravity 0.911
Stabilizer 0.01% BHT
Formula Weight 118.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material trans-β-Methylstyrene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-848-0
RTECSNumber DA8400500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.