Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Decane, MP Biomedicals™

Catalog No. ICN19512380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19512380 100 mL
ICN19512390 500 mL
MP219512396 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19512380 Supplier MP Biomedicals, Inc Supplier No. 0219512380
Add to Cart
Add to Cart

n-Decane >99%, Clear, Colorless Liquid

  • Decane has been used as a solvent in the preparation of high alkane phase emulsified suspensions (HAPES).
  • It may also be used as a solvent to suspend the CdSe/ZnS nanocrystals during absorbance measurement.
  • Internal standard in the GC analysis of oxalic, malonic, and succinic acids in biological materials.
  • Decane (n-Decane), an aliphatic hydrocarbon, is a reference fuel. Its oxidation over a range of temperature has been studied.
  • Its crystal structure has been investigated at 150K.
  • The cracking of decane can be performed using electrohydrodynamic atomization system (EHDA).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-18-5
Molecular Formula C10H22
Molecular Weight (g/mol) 142.29
InChI Key DIOQZVSQGTUSAI-UHFFFAOYSA-N
Synonym n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
PubChem CID 15600
ChEBI CHEBI:41808
IUPAC Name decane
SMILES CCCCCCCCCC

Specifications

Melting Point −30°C (Literature)
Color Colorless
Density 0.734g/cm3 at 15°C(Lit.)
Boiling Point 174°C (Literature)
Refractive Index n20/D 1.411(Lit.)
Quantity 100 mL
UN Number UN1760
Formula Weight 142.29
Percent Purity 0.996
Physical Form Liquid
Chemical Name or Material n-Decane
Show More Show Less
storageNote1 Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.