Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
n-Decane (Decane; Decyl Hydride), Reagents
Click to view available options
Quantity:
100 mL
450 mL
Specifications
Specifications
Boiling Range | 174°C |
Melting Point | -29.7°C |
Color | Colorless |
Boiling Point | 174°C |
Melting Range | -29.7°C |
Packaging | Glass Bottle |
Quantity | 450 mL |
Specific Gravity | 0.73 |
Grade | Reagent |
Physical Form | Liquid |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction