Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ N-DEX™ B6005PF Disposable Nitrile Gloves

Catalog No. 191202523E Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-120-2523E X-Large
19-120-2523A X-Small
19-120-2523B Small
19-120-2523C Medium
19-120-2523D Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-120-2523E Supplier SHOWA™ Supplier No. B6005PFXL
This item is not returnable. View return policy

N-DEX™ B6005 series powder free gloves are clean-processed and are ideal for food, utilities, pharmaceutical and electronics markets.

  • Clean-processed, powder free
  • Ambidextrous
  • Rolled cuff
  • Smooth grip
  • Made in the U.S.A.

Specifications

Certifications/Compliance FDA 21 CFR 170-199
Applications Industrial, Laboratory
AQL 4
Color Blue
Disposable Single-use
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Large
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Model B6005PFXL
Length (English) 9.5 in.
Length (Metric) 24 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.