Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-dodecane, >99%, MP Biomedicals™

Catalog No. ICN19515880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19515880 100 mL
ICN19515825 25 mL
ICN19515890 500 mL
ICN19515891 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19515880 Supplier MP Biomedicals, Inc Supplier No. 0219515880
Add to Cart
Add to Cart

Chemical Identifiers

CAS 112-40-3
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
MDL Number MFCD00008969
InChI Key SNRUBQQJIBEYMU-UHFFFAOYSA-N
Synonym n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
PubChem CID 8182
ChEBI CHEBI:28817
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Specifications

Color Colorless
Density 0.753 at 25°C
Assay Percent Range >99%
Quantity 100 mL
Formula Weight 170.34
Percent Purity 0.995
Physical Form Liquid
Chemical Name or Material N-dodecane
EINECSNumber 203-967-9
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.