Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Dodecane, MP Biomedicals™

Catalog No. MP219515896 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219515896 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219515896 Supplier MP Biomedicals, Inc Supplier No. 0219515896

missing translation for 'validMainframeMsg'

Used as a solvent, distillation chaser and scintillator component. n-Dodecane, MP Biomedicals is a liquid alkane hydrocarbon that is also used as a diluent for tributyl phosphate (TBP) in plants reprocessing.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-40-3
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
InChI Key SNRUBQQJIBEYMU-UHFFFAOYSA-N
Synonym n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
PubChem CID 8182
ChEBI CHEBI:28817
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Specifications

Melting Point -9.6°C (literature)
Density 0.753 at 25°C (water = 1) (Lit.)
Boiling Point 215°C to 217°C (literature)
Refractive Index n20/D 1.421 (Lit.)
Quantity 2.5 L
Formula Weight 170.34g/mol
Physical Form Liquid
Recommended Storage Store at Room Temperature (15–30°C). Store Desiccated.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.