Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Dodecyl-beta-D-maltopyranoside

Catalog No. p-7043285
Click to view available options
Quantity:
500 mg
1 g
5 g

A non-ionic detergent useful in membrane solubilization studies | CAS: 69227-93-6 | C24H46O11 | 510.621 g/mol

N-dodecyl-β-D-maltoside is used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research. It has also been used for the isolation of the mitochondrial cytochrome c oxidize.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-dodecyl-β-D-maltoside is used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research. It has also been used for the isolation of the mitochondrial cytochrome c oxidize.

Solubility
Soluble in water at 10% w/v

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69227-93-6
Molecular Formula C24H46O11
Molecular Weight (g/mol) 510.621
MDL Number MFCD00043012
InChI Key NLEBIOOXCVAHBD-QKMCSOCLSA-N
Synonym n-dodecyl-beta-d-maltoside, lauryl maltoside, dodecyl maltoside, dodecyl-beta-d-maltoside, n-dodecyl b-d-maltoside, unii-di107e57b4, lauryl-beta-d-maltoside, dodecyl beta-d-maltoside, dodecyl b-d-maltopyranoside, dodecyl beta-d-maltopyranoside
PubChem CID 114880
ChEBI CHEBI:43769
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O

Specifications

Melting Point 224°C to 226°C
Color White
Quantity 500 mg
Beilstein 55318
Solubility Information Soluble in water at 10% w/v
Formula Weight 510.62
Physical Form Powder
Chemical Name or Material n-Dodecyl-beta-D-maltopyranoside
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.