Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ n-Dodecyl-β-D-Maltoside Detergent, 1 g, 1 g PROMO

Catalog No. PI89902PM
Click to view available options
:
Thermo Scientific™ n-Dodecyl-β-D-Maltoside Detergent

Catalog No. PI89902PM

Supplier: Thermo Scientific™ 89902

Maltoside detergents are a class of nonionic surfactants that are commonly utilized in the field of membrane protein solubilization. Several studies have shown that these surfactants are highly effective in extracting and stabilizing membrane proteins.

Features of maltoside detergents

  • Lipid-like nonionic detergent
  • Especially useful for isolating and stabilizing hydrophobic membrane proteins
  • Preserves activity of membrane protein better than most of the detergents
  • High-purity compounds with low UV absorptivity

n-Dodecyl-β-D-Maltoside

n-Dodecyl-β-D-Maltoside (DDM) is especially useful for solubilizing membrane proteins to preserve their activity. This water-soluble nonionic detergent is most often used for the isolation of hydrophobic membrane proteins. Multiple studies have shown that n-dodecyl-β-D-maltoside is a gentle detergent that is often able to preserve protein activity better than many commonly used detergents, including NP-40, CHAPS, and octyl-β-glucoside.

Properties of DDM

  • Aggregation number: 98 (average), 70 to 140 range
  • Micelle molecular weight: 50,000 g
  • CMC: 0.17 mM (0.009%, w/v) in water ; 0.12 mM (0.006%, w/v) in 0.2 M NaCl

DDM/CHS Solution

DDM/CHS Solution (10:1) is a ready-to-use solution for solubilization of membrane proteins while maintaining structural integrity and activity. The stable pre-formulated solution contains a mixture of dodecyl-β-D-maltoside (DDM) (100 mg/mL) and cholesteryl hemisuccinate (CHS) (10 mg/mL) in deionized water with final concentrations of 10% and 1%, respectively. Typical working concentrations are 1% DDM/0.1% CHS for protein extraction and 0.1% DDM/0.01% CHS for protein purification. The mixture of nonionic detergent, DDM, with dual hydrophobic/hydrophilic properties and anionic detergent, CHS, aid in the stabilization of large unilamellar vesicles and is suitable for protein solubilization and stabilization for single particle cryo-EM analysis.

Additional properties of CHS

  • Molecular formula:;C31H50O4 · C4H11NO3
  • Molecular weight:;607.9 g/mol
  • Detergent: ionizable anionic
  • Function:; stabilizes large unilamellar vesicles

Lauryl Maltose Neopentyl Glycol

Lauryl Maltose Neopentyl Glycol (LMNG) is an amphiphilic detergent ideally suited to improve solubilization of integral membrane proteins while maintaining structural integrity and activity. LMNG has two linked hydrophobic chains of equal length and two hydrophilic maltoside groups to better mimic the integral membrane protein environment. Micelles are known to aid in solubilization of membrane proteins while preserving structure and function, including G-protein coupled receptors (GPCRs). LMNG is suitable for protein solubilization and stabilization for single particle cryo-EM analysis.

Properties of LMNG

  • Chemical name:;2,2-didecylpropane-1,3-bis-β-D-maltopyranoside
  • Molecular weight:;1005.19 g/mol
  • CMC:;(H2O) ∼ 0.01 mM, (0.001%)
  • Purity:;≥98% all anomers (by HPLC analysis)

LMNG/CHS Solution

LMNG/CHS Solution (10:1) is a ready-to-use formulation for the solubilization of integral membrane proteins while maintaining structural integrity and activity. The stable pre-formulated solution contains a mixture of lauryl maltose neopentyl glycol (LMNG) (5%) and cholesteryl hemisuccinate (CHS) (0.5%) in deionized water. Typical working concentrations for protein extraction are 1% LMNG/0.1% CHS and 0.01% LMNG/0.001% CHS for protein purification and structure determination. LMNG/CHS micelles aid in solubilization of membrane proteins while preserving structure and function, including G-protein coupled receptors (GPCRs), and is suitable for protein solubilization and stabilization for single particle cryo-EM analysis.

n-Undecyl-β-D-Maltopyranoside

n-Undecyl-β-D-Maltopyranoside (UDM) has an undecyl (C11) alkyl chain attached to the maltose headgroup. The longer alkyl chain length of UDM contributes to its enhanced solubilization efficiency for certain membrane proteins. The increased hydrophobicity of the undecyl chain may facilitate stronger interactions with hydrophobic regions of membrane proteins, leading to improved solubilization and stabilization. The longer undecyl chain length in UDM may also result in a higher CMC compared to maltosides with shorter alkyl chains. This can affect the formation and stability of micelles in solution.

Properties of UDM

  • CMC: (H2O) ~0.59 mM(1) (0.029%)
  • Aggregation number: ~71 (100 mM NaCl, 20 mM HEPES pH 7.5)
  • Purity: ≥99% β+α (by HPLC analysis)

n-Nonyl-Beta-Maltoside

n-Nonyl-Beta-Maltoside (NM) has a nonyl (C9) alkyl chain attached to the maltose headgroup. This specific chain length provides NM with distinct solubilization and stabilization properties compared to other maltosides with different alkyl chain lengths, such as octyl or decyl maltoside. The specific alkyl chain length of NM influences its micelle properties and contributes to a relatively low CMC. This property allows for the formation of stable micelles at lower concentrations, important for maintaining the stability and integrity of membrane proteins during solubilization and subsequent experiments.

Properties of NM

  • CMC: (H2O) ~ 6 mM (0.28%)
  • Aggregation number: ~55 (100 mM NaCl, 20 mM HEPES pH 7.5)
  • Purity: ≥99% β+α (by HPLC analysis)

n-Octyl-β-D-Maltopyranoside

n-Octyl-β-D-Maltopyranoside (OM) has an octyl (C8) alkyl chain attached to the maltose headgroup. The specific alkyl chain length of OM sets it apart from other maltosides with different alkyl chain lengths, such as nonyl or decyl maltoside. The octyl chain length in OM contributes to a relatively low CMC, allowing for the formation of stable micelles at lower concentrations. This property is important for maintaining the stability and integrity of membrane proteins during solubilization and subsequent experiments. OM is versatile and finds applications in various areas of membrane protein research, protein crystallization, and biochemical studies. It can be used in a wide range of experimental conditions, buffer systems, and pH ranges. OM offers flexibility and compatibility with different experimental setups.

Properties of OM

  • CMC: (100 mM NaCl, 20 mM HEPES pH 7.5) ~ 19.5 mM(1) (0.89%)
  • Aggregation number: ~47 (100 mM NaCl, 20 mM HEPES pH 7.5)
  • Purity: ≥99% β+α (by HPLC analysis)

Specifications

Product Type Detergent
Content And Storage Store below -20°C.
Form Powder
Reagent Type n-Dodecyl-β-D-Maltoside
Quantity 1 g
Molecular Weight (g/mol) 510.6 g/mol

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.