Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Dodecyl methyl sulfide, Thermo Scientific™

Catalog No. AA1487714
Click to view available options
Quantity:
5 g
25 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3698-89-3
Molecular Formula C13H28S
Molecular Weight (g/mol) 216.43
MDL Number MFCD00015073
InChI Key KJWHJDGMOQJLGF-UHFFFAOYSA-N
Synonym dodecyl methyl sulfide, 2-thiatetradecane, 1-methylsulfanyl dodecane, methylthiododecane, methyl lauryl sulfide, dodecane, 1-methylthio, methyl n-dodecyl sulfide, dodecyl methyl sulphide, sulfide, dodecyl methyl, unii-77bo1dv08u
PubChem CID 77287
IUPAC Name 1-methylsulfanyldodecane
SMILES CCCCCCCCCCCCSC

Specifications

Boiling Point 163°C to 165°C (19 mmHg)
Odor Sulphurous
Refractive Index 1.4608
Linear Formula CH3(CH2)11SCH3
Quantity 25 g
Formula Weight 216.41
Physical Form Liquid
Chemical Name or Material n-Dodecyl methyl sulfide
EINECSNumber 223-028-7
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.