Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Dodecyl Sulfide 93%, Thermo Scientific™

Catalog No. AC117681000
Click to view available options
:
100g; Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2469-45-6
CAS Min % 91.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 93%
Packaging Glass bottle
Molecular Formula C24H50S
Linear Formula [CH3(CH2)11]2S
Quantity 100g
MDL Number MFCD00008975
Synonym dodecyl sulfide, didodecyl sulfide, dilauryl sulfide, n-dodecyl sulfide, dodecane, 1,1'-thiobis, bis dodecyl sulfide, bis n-dodecyl sulfide, unii-m8cf2nuw66, m8cf2nuw66, dodecylthiododecane
InChI Key UPYPTOCXMIWHSG-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCSCCCCCCCCCCCC
IUPAC Name 1-dodecylsulfanyldodecane
Molecular Weight (g/mol) 370.724
PubChem CID 72875
Formula Weight 370.71
Percent Purity ≥91%
Chemical Name or Material n-Dodecyl sulfide, 93%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-587-1
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.