Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Dodecylacrylamide 97.0+%, TCI America™
SDP

Catalog No. D331625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D331625G 25 g
D33165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D331625G Supplier TCI America Supplier No. D331625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1506-53-2
Molecular Formula C15H29NO
Molecular Weight (g/mol) 239.40
MDL Number MFCD08276361
InChI Key XQPVIMDDIXCFFS-UHFFFAOYSA-N
Synonym n-dodecylacrylamide, 2-propenamide,n-dodecyl, acmc-209d4r
PubChem CID 73929
IUPAC Name N-dodecylprop-2-enamide
SMILES CCCCCCCCCCCCNC(=O)C=C

Specifications

Melting Point 58°C
Color White
Boiling Point 155°C
Quantity 25 g
Formula Weight 239.40
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Dodecylacrylamide
EINECSNumber (2)-1019
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.