Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethyl-o-crotonotoluidide 95.0+%, TCI America™
SDP

Catalog No. E081425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E081425G 25 g
E08145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E081425G Supplier TCI America Supplier No. E081425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 124236-29-9
Molecular Formula C13H17NO
Molecular Weight (g/mol) 203.285
MDL Number MFCD00026989
InChI Key DNTGGZPQPQTDQF-XBXARRHUSA-N
Synonym trans-N-Ethyl-N-(o-tolyl)-2-butenamide, N-Crotonyl-N-ethyl-o-toluidine
PubChem CID 688020
IUPAC Name (E)-N-ethyl-N-(2-methylphenyl)but-2-enamide
SMILES CCN(C1=CC=CC=C1C)C(=O)C=CC

Specifications

Color Yellow
Boiling Point 155°C
Quantity 25 g
Formula Weight 203.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Ethyl-o-crotonotoluidide
EINECSNumber (9)-0214
RTECSNumber GQ7000000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.