Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethylaniline, 98%

Catalog No. AC118045000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle

CAS: 103-69-5 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-69-5
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00009025
InChI Key OJGMBLNIHDZDGS-UHFFFAOYSA-N
Synonym ethylaniline, n-ethyl aniline, benzenamine, n-ethyl, anilinoethane, n-ethylbenzenamine, ethylphenylamine, n-ethyl-n-phenylamine, aniline, n-ethyl, n-ethylaminobenzene, aethylanilin
PubChem CID 7670
ChEBI CHEBI:34870
IUPAC Name N-ethylaniline
SMILES CCNC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -63°C
Color Brown to Yellow
Density 0.9630g/mL
Boiling Point 205°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5530 to 1.555
Linear Formula C6H5NHC2H5
Quantity 500 g
Beilstein 12, 159
Specific Gravity 0.963
Merck Index 15, 3819
Solubility Information Solubility in water: 50g/L water (20°C). Other solubilities: soluble in alcohol,ether and many other organic,solvents
Formula Weight 121.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Ethylaniline, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-135-5
RTECSNumber BX9780000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.