Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethylethylenediamine, 98%

Catalog No. AAB2357714
Click to view available options
Quantity:
5 g
25 g

CAS: 110-72-5 | C4H12N2 | 88.154 g/mol

N-Ethylethylenediamine is a chelating agent used in mutagenesis research studies. It undergoes condensation with 2-hydroxy-3-methoxybenzaldehyde. It also forms organometallic ruthenium(II) complexes that inhibit the growth in human ovarian cancer cell lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Ethylethylenediamine is a chelating agent used in mutagenesis research studies. It undergoes condensation with 2-hydroxy-3-methoxybenzaldehyde. It also forms organometallic ruthenium(II) complexes that inhibit the growth in human ovarian cancer cell lines.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-72-5
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.154
MDL Number MFCD00008166
InChI Key SCZVXVGZMZRGRU-UHFFFAOYSA-N
Synonym n-ethylethylenediamine, 1,2-ethanediamine, n-ethyl, 2-aminoethyl ethyl amine, 2-ethylaminoethylamine, ethylenediamine, n-ethyl, ethylaminoethylamine, unii-941mvd708n, 1,2-ethanediamine, n1-ethyl, n-ethyl ethylenediamine
PubChem CID 66071
IUPAC Name N'-ethylethane-1,2-diamine
SMILES CCNCCN

Specifications

Density 0.837
Boiling Point 128°C to 130°C
Flash Point 34°C (93°F)
Refractive Index 1.44
Quantity 25 g
UN Number UN2733
Beilstein 1098272
Sensitivity Air Sensitive
Formula Weight 88.15
Percent Purity 98%
Chemical Name or Material N-Ethylethylenediamine
Show More Show Less
Hazard Category H225-H314
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 203-795-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.