Learn More
N-Ethylmaleimide, 99+%
Description
N-Ethylmaleimide is a protein thiol modifier which inhibits apoptotic DNA fragmentation. It is a sulfhydryl reagent and finds application in experimental biochemical studies as well as in enzymology. It is involved in the modification of cysteine residues in proteins and peptides. It acts as a Michael acceptor and reacts with nucelophiles like thiols. It is employed in the inhibition of Mg2+ dependent internucleosomal DNA fragmentation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 43°C to 46°C |
Color | Cream to White |
Boiling Point | 209°C to 211°C |
Flash Point | 73°C (163°F) |
Odor | Pungent |
Quantity | 25 g |
UN Number | UN2928 |
Beilstein | 112448 |
Sensitivity | Light sensitive |
Merck Index | 14,3822 |
Show More |
Safety and Handling
Hazard Category | H300-H311-H314-H317-H335 |
Hazard Statement | GHS H Statement H300-H311-H314-H318-H317; Fatal if swallowed. Toxic in contact with skin. Causes severe skin burns and eye damage. Causes serious eye damage. May cause an allergic skin reaction. |
Precautionary Statement | P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S. |
EINECSNumber | 204-892-4 |
RTECSNumber | UX9625000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.