Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-2-cyclopentyl-L-glycine, 95%

Click to view available options
Quantity:
250mg
1g
5g
220497-61-0
C22H23NO4
365.429
MFCD01311751
JGWHUIQSEKGLAQ-FQEVSTJZSA-N
fmoc-cpg-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclopentylacetic acid,fmoc-cyclopentyl-gly-oh,s-cyclopentyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid,fmoc-l-cpg-oh,ambotzfaa1360,fmoc-l-cyclopentyl-glycine,n-fmoc-alpha-cyclopentylglycine,fmoc-s-2-cyclopentylglycine,n-alpha-9-fluorenylmethyloxycarbonyl-l-beta-cyclopentylglycine
7006544
(2S)-2-cyclopentyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid
C1CCC(C1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
220497-61-0
C22H23NO4
365.429
MFCD01311751
JGWHUIQSEKGLAQ-FQEVSTJZSA-N
fmoc-cpg-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclopentylacetic acid,fmoc-cyclopentyl-gly-oh,s-cyclopentyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid,fmoc-l-cpg-oh,ambotzfaa1360,fmoc-l-cyclopentyl-glycine,n-fmoc-alpha-cyclopentylglycine,fmoc-s-2-cyclopentylglycine,n-alpha-9-fluorenylmethyloxycarbonyl-l-beta-cyclopentylglycine
7006544
(2S)-2-cyclopentyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid
C1CCC(C1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Formula Weight 379.46
Physical Form Powder
Percent Purity 95%
Color White
Quantity 1g
Chemical Name or Material N-Fmoc-2-cyclopentyl-L-glycine

storageNote1 : -30° to -10°C