Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-2-methyl-D-phenylalanine, 95%, Thermo Scientific™

Catalog No. AAH6518806
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352351-63-4
Molecular Formula C25H23NO4
Molecular Weight (g/mol) 401.462
MDL Number MFCD00671404
InChI Key GYFMRMRGTNDDAT-HSZRJFAPSA-N
Synonym fmoc-2-methyl-d-phenylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-o-tolyl propanoic acid, fmoc-d-2-methylphenylalanine, fmoc-d-2-methylphe, fmoc-d-phe 2-me-oh, fmoc-d-2-me-phe-oh, fmoc-l-phe 2-me-oh, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-2-methylphenyl propanoic acid, fmoc-o-me-d-phe-oh, fmoc-2-methy-d-phenylalanine
PubChem CID 7129821
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-methylphenyl)propanoic acid
SMILES CC1=CC=CC=C1CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point 110°C to 116°C
Color White
Beilstein 9157614
Quantity 5 g
Formula Weight 401.46
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-2-methyl-D-phenylalanine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.