Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-3-(4-pyridyl)-L-alanine, 95%

Catalog No. AAH5217203
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 169555-95-7 | C23H20N2O4 | 388.423 g/mol

Fmoc-(4-pyridyl)-Ala-OH is potentially useful for solid phase peptide synthesis techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fmoc-∫-(4-pyridyl)-Ala-OH is potentially useful for solid phase peptide synthesis techniques.

Solubility
Soluble in 0.1g in 4mL DMF(clearly soluble).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169555-95-7
Molecular Formula C23H20N2O4
Molecular Weight (g/mol) 388.423
MDL Number MFCD00672566
InChI Key SCSSXJVRZMQUKA-NRFANRHFSA-N
Synonym fmoc-l-4-pyridylalanine, fmoc-4-pal-oh, fmoc-3-4-pyridyl-l-alanine, fmoc-ala 4-pyridyl-oh, fmoc-beta-4-pyridyl-ala-oh, n-fmoc-3-4-pyridyl-l-alanine, fmoc-l-3-4-pyridyl-alanine, fmoc-4'-pyridyl-l-ala, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-4-yl propanoic acid, fmoc-3-4-pyridyl-d-alanine
PubChem CID 978322
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=NC=C4)C(=O)O

Specifications

Quantity 1 g
Beilstein 7316905
Solubility Information Soluble in 0.1g in 4mL DMF(clearly soluble).
Formula Weight 388.42
Percent Purity 95%
Chemical Name or Material N-Fmoc-3-(4-pyridyl)-L-alanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.