Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-3-nitro-D-phenylalanine, 95%

Click to view available options
Quantity:
250mg
1g
5g
478183-71-0
C24H20N2O6
432.432
MFCD01317715
UDIZJKKIJYRJIN-JOCHJYFZSA-N
fmoc-3-nitro-d-phenylalanine,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-3-nitrophenyl propanoic acid,fmoc-d-3-nitrophenylalanine,fmoc-d-3-no2-phe-oh,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-3-nitrophenyl propanoic acid,fmoc-l-phe 3-no2-oh,fmoc-d-3-nitrophe,n-9h-fluoren-9-yl methoxy carbonyl-3-nitro-d-phenylalanine,2r-2 9h-fluoren-9-yl methoxy carbonyl amino-3-3-nitrophenyl propanoic acid
7006635
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(3-nitrophenyl)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC(=CC=C4)[N+](=O)[O-])C(=O)O
478183-71-0
C24H20N2O6
432.432
MFCD01317715
UDIZJKKIJYRJIN-JOCHJYFZSA-N
fmoc-3-nitro-d-phenylalanine,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-3-nitrophenyl propanoic acid,fmoc-d-3-nitrophenylalanine,fmoc-d-3-no2-phe-oh,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-3-nitrophenyl propanoic acid,fmoc-l-phe 3-no2-oh,fmoc-d-3-nitrophe,n-9h-fluoren-9-yl methoxy carbonyl-3-nitro-d-phenylalanine,2r-2 9h-fluoren-9-yl methoxy carbonyl amino-3-3-nitrophenyl propanoic acid
7006635
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(3-nitrophenyl)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC(=CC=C4)[N+](=O)[O-])C(=O)O
Formula Weight 432.43
Physical Form Powder
Percent Purity 95%
Color White
Melting Point 151°C to 157°C
Quantity 5g
Chemical Name or Material N-Fmoc-3-nitro-D-phenylalanine

storageNote1 : 2° to 8°C