Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-D-tyrosine, 97%

Catalog No. AAH6674003
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 112883-29-1 | C24H21NO5 | 403.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112883-29-1
Molecular Formula C24H21NO5
Molecular Weight (g/mol) 403.43
MDL Number MFCD00171384
InChI Key SWZCTMTWRHEBIN-ANBDAQEENA-N
Synonym fmoc-d-tyrosine, fmoc-d-tyr-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-hydroxyphenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-hydroxyphenyl propanoic acid, n-fmoc-d-tyr oh, fmoc-d-phe 4-oh-oh, n-9h-fluorene-9-ylmethoxycarbonyl-d-tyrosine, n-alpha-9-fluorenylmethoxycarbonyl-d-tyrosine, r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-4-hydroxy-phenyl-propionic acid
PubChem CID 6957988
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-hydroxyphenyl)propanoic acid
SMILES OC(=O)[C@@H](CC1=CC=C(O)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 175°C to 180°C
Color White
Quantity 1 g
Formula Weight 403.43
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Fmoc-D-tyrosine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.