Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-FMOC-L-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid, 99%, Thermo Scientific™

Catalog No. AC301922500
Click to view available options
:
250mg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 136030-33-6
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C25H21NO4
Quantity 250mg
MDL Number MFCD00144368
Synonym fmoc-tic-oh, fmoc-l-tic, fmoc-l-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-2-9h-fluoren-9-yl methoxy carbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-+-2-9-fluorenylmethoxycarbonyl-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, n-fmoc-l-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3s-2-9h-fluoren-9-ylmethoxycarbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid, 3s-2-9h-fluoren-9-ylmethoxy carbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid, fmoc-tic, pubchem6028
InChI Key LIRBCUNCXDZOOU-QHCPKHFHSA-N
SMILES C1C(N(CC2=CC=CC=C21)C(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)C(=O)O
IUPAC Name (3S)-2-(9H-fluoren-9-ylmethoxycarbonyl)-3,4-dihydro-1H-isoquinoline-3-carboxylic acid
Molecular Weight (g/mol) 399.446
PubChem CID 978341
Formula Weight 399.45
Percent Purity ≥98.5%
Chemical Name or Material N-FMOC-L-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.