Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester is used as organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester is used as organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester is used as organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 115°C to 120°C |
Quantity | 1 g |
Beilstein | 8019724 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 439.51 |
Percent Purity | 95% |
Chemical Name or Material | N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester |
Safety and Handling
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction