Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-glutamic acid 5-tert-butyl ester pentafluorophenyl ester, 98%

Click to view available options
Quantity:
5g
25g
86061-04-3
C30H26F5NO6
591.531
MFCD00065647
AIDYQYOPUBOMTR-FQEVSTJZSA-N
fmoc-glu otbu-opfp,5-tert-butyl 1-pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate,fmoc-glu otbu-opfp,fmoc-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester,n-fmoc-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester,5-tert-butyl1-pentafluorophenyln-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate,n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 1-pentafluorophenyl 5-tert-butyl ester,1-tert-butyl 2,3,4,5,6-pentafluorophenyl 4s-4-9h-fluoren-9-ylmethoxy carbonyl amino pentanedioate,n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester,n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-t-butyl ester pentafluorphenyl ester
11227121
5-O-tert-butyl 1-O-(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioate
CC(C)(C)OC(=O)CCC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
86061-04-3
C30H26F5NO6
591.531
MFCD00065647
AIDYQYOPUBOMTR-FQEVSTJZSA-N
fmoc-glu otbu-opfp,5-tert-butyl 1-pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate,fmoc-glu otbu-opfp,fmoc-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester,n-fmoc-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester,5-tert-butyl1-pentafluorophenyln-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate,n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 1-pentafluorophenyl 5-tert-butyl ester,1-tert-butyl 2,3,4,5,6-pentafluorophenyl 4s-4-9h-fluoren-9-ylmethoxy carbonyl amino pentanedioate,n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester,n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-t-butyl ester pentafluorphenyl ester
11227121
5-O-tert-butyl 1-O-(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioate
CC(C)(C)OC(=O)CCC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Beilstein 3642680
Formula Weight 591.53
Physical Form Solid
Percent Purity 98%
Quantity 25g
Chemical Name or Material N-Fmoc-L-glutamic acid 5-tert-butyl ester pentafluorophenyl ester

storageNote1 : 2° to 8°C