Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-prolyl-L-proline, 95%

Click to view available options
Quantity:
1g
5g
129223-22-9
C25H26N2O5
434.492
MFCD00237673
VRAQFWSWKRNOGU-VXKWHMMOSA-N
fmoc-pro-pro-oh,s-1-s-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic acid,l-proline, 1-9h-fluoren-9-ylmethoxy carbonyl-l-prolyl,2s-1-2s-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic acid,s-1-s-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic aci,fmoc-pro-pro,fmoc-prolinyl-proline,fmoc-prolylproline-oh,fmoc-l-pro-l-pro-oh,fmoc-prolinyl-proline s,s
11133845
(2S)-1-[(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)pyrrolidine-2-carbonyl]pyrrolidine-2-carboxylic acid
C1CC(N(C1)C(=O)C2CCCN2C(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)C(=O)O
129223-22-9
C25H26N2O5
434.492
MFCD00237673
VRAQFWSWKRNOGU-VXKWHMMOSA-N
fmoc-pro-pro-oh,s-1-s-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic acid,l-proline, 1-9h-fluoren-9-ylmethoxy carbonyl-l-prolyl,2s-1-2s-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic acid,s-1-s-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic aci,fmoc-pro-pro,fmoc-prolinyl-proline,fmoc-prolylproline-oh,fmoc-l-pro-l-pro-oh,fmoc-prolinyl-proline s,s
11133845
(2S)-1-[(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)pyrrolidine-2-carbonyl]pyrrolidine-2-carboxylic acid
C1CC(N(C1)C(=O)C2CCCN2C(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)C(=O)O
Formula Weight 434.49
Physical Form Solid
Percent Purity 95%
Quantity 1g
Chemical Name or Material N-Fmoc-L-prolyl-L-proline

storageNote1 : -30° to -10°C