Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-valinol, 98%

Click to view available options
Quantity:
1g
5g
160885-98-3
C20H23NO3
325.408
MFCD00235961
MYMGENAMKAPEMT-LJQANCHMSA-N
fmoc-valinol,fmoc-l-valinol,n-fmoc-l-valinol,s-9h-fluoren-9-yl methyl 1-hydroxy-3-methylbutan-2-yl carbamate,9h-fluoren-9-ylmethyl n-2s-1-hydroxy-3-methylbutan-2-yl carbamate,n-fmoc-l-valinol,n-9-fluorenylmethoxycarbonyl-l-valinol,s-2-fmoc-amino-3-methyl-1-butanol,s-2-9h-fluorene-9-ylmethoxycarbonylamino-3-methyl-1-butanol,s-2-9-fluorenylmethyloxycarbonyl amino-3-methyl-1-butanol, n-alpha-9-fluorenylmethyloxycarbonyl-l-valinol
688273
9H-fluoren-9-ylmethyl N-[(2S)-1-hydroxy-3-methylbutan-2-yl]carbamate
CC(C)C(CO)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
160885-98-3
C20H23NO3
325.408
MFCD00235961
MYMGENAMKAPEMT-LJQANCHMSA-N
fmoc-valinol,fmoc-l-valinol,n-fmoc-l-valinol,s-9h-fluoren-9-yl methyl 1-hydroxy-3-methylbutan-2-yl carbamate,9h-fluoren-9-ylmethyl n-2s-1-hydroxy-3-methylbutan-2-yl carbamate,n-fmoc-l-valinol,n-9-fluorenylmethoxycarbonyl-l-valinol,s-2-fmoc-amino-3-methyl-1-butanol,s-2-9h-fluorene-9-ylmethoxycarbonylamino-3-methyl-1-butanol,s-2-9-fluorenylmethyloxycarbonyl amino-3-methyl-1-butanol, n-alpha-9-fluorenylmethyloxycarbonyl-l-valinol
688273
9H-fluoren-9-ylmethyl N-[(2S)-1-hydroxy-3-methylbutan-2-yl]carbamate
CC(C)C(CO)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Quantity 1g
Formula Weight 325.41
Physical Form Solid
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-valinol

storageNote1 : -30° to -10°C