Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-S-trityl-L-cysteine, 95%

Catalog No. p-7029851
Click to view available options
Quantity:
5 g
25 g

CAS: 103213-32-7 | C37H30NO4S | 584.71 g/mol

It is potentially useful for proteomics studies, and solid phase peptide synthesis techniques. Cysteine is versatile amino acid involved with many biological processes, including the formation of disulfide bonds - a critical component of protein structure. This compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is potentially useful for proteomics studies, and solid phase peptide synthesis techniques. Cysteine is versatile amino acid involved with many biological processes, including the formation of disulfide bonds - a critical component of protein structure. This compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function.

Solubility
Insoluble in water. Soluble in most organic solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103213-32-7
Molecular Formula C37H30NO4S
Molecular Weight (g/mol) 584.71
MDL Number MFCD00038538
InChI Key KLBPUVPNPAJWHZ-UMSFTDKQSA-M
Synonym fmoc-cys trt-oh, fmoc-s-trityl-l-cysteine, fmoc-l-cys trt-oh, n-fmoc-s-trityl-l-cysteine, fmoc-cys trt, n alpha-fluorenylmethyloxycarbonyl-s-tritylcysteine, r-9h-fluoren-9-yl methyl 1-hydroxy-3-tritylthio propan-2-yl carbamate, fmoc-s-trityl-cys, n-9h-fluoren-9-ylmethoxy carbonyl-s-triphenylmethyl-l-cysteine, l-cysteine, n-9h-fluoren-9-ylmethoxy carbonyl-s-triphenylmethyl
PubChem CID 128239
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-tritylsulfanylpropanoic acid
SMILES [O-]C(=O)[C@H](CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 171°C to 173°C
Quantity 5 g
Solubility Information Insoluble in water. Soluble in most organic solvents.
Optical Rotation +17° (c=1 in THF)
Formula Weight 585.71
Percent Purity 95%
Chemical Name or Material N-Fmoc-S-trityl-L-cysteine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.