Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-S-trityl-L-cysteine pentafluorophenyl ester, 98%

Catalog No. AAH63003MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63003MD 250 mg
AAH6300303 1 g
AAH6300306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63003MD Supplier Thermo Scientific Chemicals Supplier No. H63003MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

CAS: 115520-21-3 | C43H30F5NO4S | 751.768 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115520-21-3
Molecular Formula C43H30F5NO4S
Molecular Weight (g/mol) 751.768
MDL Number MFCD00065639
InChI Key FDAUCYDVXIPBDR-UMSFTDKQSA-N
Synonym fmoc-cys trt-opfp, r-perfluorophenyl 2-9h-fluoren-9-yl methoxy carbonyl amino-3-tritylthio propanoate, fmoc-s-trityl-l-cysteine pentafluorophenyl ester, 2,3,4,5,6-pentafluorophenyl 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-triphenylmethyl sulfanyl propanoate, fmoc-cys trt-opfp hplc, n, a-fmoc-s-trityl-l-cysteine pentafluorophenyl ester, nalpha-fmoc-s-trityl-l-cysteine pentafluorophenyl ester, n alpha-fmoc-s-trityl-l-cysteine penta-fluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-s-trityl-l-cysteine pentafluorphenyl ester, pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-s-trityl-l-cysteinate
PubChem CID 11050947
IUPAC Name (2,3,4,5,6-pentafluorophenyl) (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-tritylsulfanylpropanoate
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)SCC(C(=O)OC4=C(C(=C(C(=C4F)F)F)F)F)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57

Specifications

Beilstein 4225739
Quantity 250 mg
Formula Weight 751.76
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Fmoc-S-trityl-L-cysteine pentafluorophenyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.