Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Gamma-Maleimidobutyryloxy)Succinimide, PROMO, Thermo Scientific Chemicals

Catalog No. J61850MCPM
Click to view available options
Quantity:
100 mg
  • A heterobifunctional cross-linking reagent with amine and sulfhydryl reactivity
  • Useful in enzyme immunoassays
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80307-12-6
Molecular Formula C12H12N2O6
Molecular Weight (g/mol) 280.24 g/mol
MDL Number MFCD00036817
InChI Key PVGATNRYUYNBHO-UHFFFAOYSA-N
Synonym gmbs, 4-maleimidobutyric acid n-hydroxysuccinimide ester, 4-maleimidobutyric acid n-succinimidyl ester, n-gamma-maleimidobutyryloxy succinimide, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl butanoate, n-4-maleimidobutyryloxy succinimide, n-succinimidyl 4-maleimidobutyrate, n-succinimidyl-4-maleimidobutyrate, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxopyrrol-1-yl butanoate
PubChem CID 133440
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-(2,5-dioxopyrrol-1-yl)butanoate
SMILES O=C(CCCN1C(=O)C=CC1=O)ON1C(=O)CCC1=O

Specifications

Melting Point 130°C to 132°C
Color White
Quantity 100 mg
Beilstein 6427689
Physical Form Powder
Chemical Name or Material N-(gamma-Maleimidobutyryloxy)succinimide
TSCA No
Recommended Storage Store at -20°C

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.