Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Glycyl-L-valine, 99%, Thermo Scientific™

Catalog No. AC277560050
Click to view available options
:
5g; Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1963-21-9
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C7H14N2O3
Infrared Spectrum Authentic
MDL Number MFCD00066046
Loss on Drying 0.2% max.
Packaging Glass bottle
Synonym glycyl-l-valine, h-gly-val-oh, gly-val, s-2-2-aminoacetamido-3-methylbutanoic acid, n-glycyl-l-valine, gly-l-val, valine, n-glycyl-, l, chembl56035, glycyl-valine, gly-d-val
InChI Key STKYPAFSDFAEPH-LURJTMIESA-N
SMILES CC(C)C(C(=O)O)NC(=O)CN
IUPAC Name (2S)-2-[(2-aminoacetyl)amino]-3-methylbutanoic acid
Specific Rotation -20° to -22° (20°C, 589nm) (c=2, H2O)
Molecular Weight (g/mol) 174.2
PubChem CID 2724807
ChEBI CHEBI:73922
Transmittance 95% min. (c=2, H2O, 430nm) 1cm cell
Formula Weight 174.2
Percent Purity ≥98.5%
Sulfated Ash 0.1% max.
Specific Rotation Condition - 21.00 (20.00°C c=2,water)
Quantity 5 g
Chemical Name or Material N-Glycyl-L-valine, 99%
Show More Show Less
EINECSNumber 217-806-5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.